Learn More
Medchemexpress LLC N,N-dihexyl-2-(4-fluorophenyl)indole-3-acetamide | 142720-24-9 | MFCD00467704 | 100.0% | 436.60 g/mol | C28H37FN2O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101059100MG
Encompass_Preferred
FGIN 1-27 is an indoleacetamide research compound that acts as a high-affinity agonist of the translocator protein (TSPO), also known as the peripheral benzodiazepine receptor (PBR). It is provided as a purified solid for preclinical in vitro and in vivo studies of mitochondrial function, neurosteroidogenesis, and TSPO-mediated pharmacology.
  • High affinity TSPO (PBR) ligand with low-nanomolar activity.
  • Indoleacetamide scaffold suitable for receptor pharmacology studies.
  • High purity (≈99.95%) for reproducible experimental results.
  • Supplied as a stable powder for storage and handling.
  • Readily soluble in DMSO; example in vivo formulations provided.
  • Suitable for both in vitro assays and in vivo pharmacology.
Catalog No. 50-002-12941