Learn More
Medchemexpress LLC N,N-dimethylglycine (standard) | 1118-68-9 | MFCD00004283 | 103.12 g/mol | C4H9NO2 | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0511R100MG
Encompass_Preferred
N N-Dimethylglycine (Dimethylglycine) (Standard) is the analytical standard of N N-Dimethylglycine (HY-Y0511) This product is intended for research and analytical applications N N-Dimethylglycine (Dimethylglycine) a natural N-methylated glycine is a nutrient supplement and acts as an NMDAR glycine site partial agonist N N-Dimethylglycine is a methyl donor that can improve immunity act as an antioxidant to prevent oxidative stress and scavenge excess free radicals N N-Dimethylglycine has antidepressant-like and surfactant effects
Catalog No. 50-002-68922