Learn More
Medchemexpress LLC N,N'-dimethylthiourea | 534-13-4 | MFCD00004923 | 99.9% | 104.17 g/mol | C3H8N2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW02795110MM/1ML
Encompass_Preferred
N,N'-Dimethylthiourea (C3H8N2S, MW 104.17 g/mol) is a thiourea derivative used in biochemical research as a reactive oxygen species (ROS) scavenger and in thiourea chemistry. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO, suitable for analytical, cellular, and synthetic applications.
  • Used as a reactive oxygen species scavenger in biochemical assays.
  • Supplied as a 10 mM solution in DMSO for ready-to-use experiments.
  • Available as solid in multiple sizes for synthesis and formulation.
  • High purity with HPLC assay of 99.93%.
  • Stable molecular profile: formula C3H8N2S, molecular weight 104.17 g/mol.
Catalog No. 50-003-01275