Learn More
Cayman Chemical N N-DIphenthylurea 5mg
SDP
Supplier:  Cayman Chemical 335805 MG
Encompass_Preferred
A bacterial metabolite with diverse biological activities; inhibits Streptomyces chitin synthase 1 (IC50 = 14 UG/ml); active against C. neoformans and R. solani (MIC = 50 UG/ml for both); enhances insulin-induced adipocyte differentiation in 3T3-L1 and C3H/10T1/2 cells at 30 and 100 µM
Catalog No. 50-295-0455