Learn More
Medchemexpress LLC N,N,N',N'-tetracyclohexyl-1,2-phenylenedioxydiacetamide | 81686-22-8 | MFCD00077941 | 99.9% | 552.79 g/mol | C34H52N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1011095MG
Encompass_Preferred
Sodium ionophore III is a sodium-selective ion carrier used to prepare sodium-selective polymeric membrane electrodes and to assay sodium activity in blood, plasma, serum, and other aqueous samples. It is supplied as a high-purity solid for research and analytical applications.
  • High sodium selectivity for use in ion-selective electrodes.
  • Suitable for assays of blood, plasma, serum, and aqueous samples.
  • High purity (~99.9%) for analytical applications.
  • Supplied as a solid powder in small laboratory pack sizes.
  • Molecular formula C34H52N2O4; molecular weight ≈552.8 g/mol.
Catalog No. 50-002-11690