Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N,N',N'-tetrakis(2-hydroxyethyl)ethylenediamine | 140-07-8 | MFCD00002854 | 99.9% | 236.31 g/mol | C10H24N2O4 | 250 G
SDP

Supplier:  Medchemexpress LLC 250GHYW047709

Encompass_Preferred

N,N,N',N'-tetrakis(2-hydroxyethyl)ethylenediamine is a tetradentate amino alcohol ligand supplied as a high-purity research chemical for coordination chemistry and biochemical applications. It functions as a chelating reagent with reported cytotoxic activity in cell assays and is provided with stability and storage guidance for laboratory use.

  • Tetradentate amine ligand for chelation and complexation
  • High purity: 99.89%
  • CAS number: 140-07-8
  • Molecular weight: 236.31 g/mol
  • Available as a 250 G solid package
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)
  • Applications: coordination chemistry, organic synthesis, biochemical research

Catalog No. 50-002-08647


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.