Learn More
Medchemexpress LLC N,N,N',N'-tetrakis(pyridin-2-ylmethyl)ethane-1,2-diamine | 16858-02-9 | MFCD00036918 | 424.54 g/mol | C26H28N6 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY100202200MG
Encompass_Preferred
TPEN (also called TPEDA) is a cell-permeable heavy-metal chelator used in biochemical and cellular research to selectively bind zinc ions. It is commonly used to probe zinc-dependent biological processes, induce oxidative stress, and study apoptosis and DNA damage in vitro. The compound is supplied as a powder with specified cold-storage recommendations to preserve stability.
  • cell-permeable heavy-metal chelator useful for zinc depletion studies.
  • higher affinity for Zn2+ than for Mg2+ or Ca2+.
  • induces intracellular reactive oxygen species and DNA damage in cell models.
  • useful for studying apoptosis and metal-dependent cellular pathways.
  • supplied as a powder with established cold-storage conditions to maintain stability.
Catalog No. 50-002-15257