Learn More
Medchemexpress LLC N, N, N′, N′-tetramethyl (succinimido) uronium tetrafluoroborate | 105832-38-0 | ≥99.0% | 301.05 | 100 G
Supplier: Medchemexpress LLC HYW011412100G
TSTU is a coupling reagent utilized in peptide chemistry. It facilitates the conversion of carboxylic acids into their corresponding N-hydroxysuccinimidyl (NHS) esters. TSTU is suitable for the in situ formation of succinimide esters in mixed aqueous/organic media for amide bond formation reactions.
- Can be applied for attaching labels to peptides, proteins, and other molecules.