Learn More
Medchemexpress LLC N N N-Trimethylpropa 10g
SDP
Supplier:  Medchemexpress LLC HYW09993010G
Encompass_Preferred
N N N-Trimethylpropan-1-aminium bromide is a quaternary ammonium compound belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis facilitating the transfer of reactants between immiscible phases It can also be used as a surfactant corrosion inhibitor or antimicrobial Due to its unique chemical properties N N N-Trimethylpropan-1-aminium bromide has applications in various industries such as pharmaceuticals agrochemicals and materials science
Catalog No. 50-002-04920