Learn More
Cayman Chemical N NDImethylcathInonhydroc 10mg
SDP
Supplier:  Cayman Chemical 900114410 MG
Encompass_Preferred
A psychotropic compound of the phenethylamine and cathinone classes; slightly less potent than methcathinone and essentially equipotent with cathinone, in behavioral studies involving rats; intended for forensic purposes
Catalog No. 50-289-4335