Learn More
Cayman Chemical N NDIpropyldopamInhydrobr 10mg
SDP
Supplier:  Cayman Chemical 2893710 MG
Encompass_Preferred
A dopamine receptor agonist; decreases DOPA levels in the limbic forebrain and striatum of reserpinized rats (ED50s = 25 and 20 µmol/kg, respectively); reduces HVA levels in rat striatum at 80 µmol/kg; reduces spontaneous locomotor activity in mice at 0.5-16 mg/kg, an effect that can be reversed by spiroperidol
Catalog No. 50-293-8424