Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(NHS ester-PEG2)-N-bis(PEG3-azide) | 99.9% | 704.73 | C28H48N8O13 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16486210MG

Encompass_Preferred

N-(NHS ester-PEG2)-N-bis(PEG3-azide) is a bifunctional polyethylene glycol (PEG) linker containing an N-hydroxysuccinimide (NHS) ester and two terminal azide groups. It is supplied as a high-purity reagent for bioconjugation and click chemistry applications, enabling amine-reactive coupling and subsequent azide-alkyne cycloaddition.

  • Reactive N-hydroxysuccinimide (NHS) ester for conjugation to primary amines.
  • Two terminal azide groups for copper-catalyzed or strain-promoted click chemistry.
  • Compact PEG spacer improves solubility and reduces steric hindrance.
  • High purity (99.90%) suitable for research applications.
  • Typical appearance: liquid; molecular weight 704.73 Da; formula C28H48N8O13.
  • Store sealed at -20°C, protected from moisture and light.
  • Available in small mg-scale quantities for research use.

Catalog No. 50-002-89283


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.