Learn More
Medchemexpress LLC Nω-nitro-L-arginine | 2149-70-4 | 99.8% | 50 G
SDP
Supplier:  Medchemexpress LLC HY1211550G
Encompass_Preferred
NG-nitro-L-arginine is a nitric oxide synthase (NOS) inhibitor used in biochemical and pharmacological research to probe nitric oxide signaling pathways. Supplied as a high-purity solid, it is suitable for in vitro assays and preparative studies.
  • High purity suitable for analytical and research applications.
  • Solid form for accurate weighing and formulation.
  • Acts as a reversible NOS inhibitor for biochemical assays.
  • Molecular formula C6H13N5O4; molecular weight 219.20 g·mol⁻1.
Catalog No. 50-002-31757