Learn More
Cayman Chemical N-NItroso DesloratdIn-d4 10mg
SDP
Supplier:  Cayman Chemical 3474110 MG
Encompass_Preferred
An internal standard for the quantification of N-nitroso desloratadine by GC- or LC-MS
Catalog No. 50-291-0715