Learn More
Medchemexpress LLC N-Nitrosomorpholine-d8 | 1219805-76-1 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY131123S11MG
Encompass_Preferred
N-Nitrosomorpholine-d8 is a deuterium-labeled version of N-Nitrosomorpholine, a light-sensitive nitrosamine and a strong animal carcinogen. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development, with potential impacts on pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium-labeled compound
  • Suitable for research use only
Catalog No. 50-003-40719