Learn More
Cayman Chemical N-NItrosonrnIcotIn-d4 10mg
SDP
Supplier:  Cayman Chemical 3560410 MG
Encompass_Preferred
An internal standard for the quantification of N'-nitrosonornicotine by GC-or LC-MS
Catalog No. 50-290-9910