Learn More
Cayman Chemical N-NItrosonrnIcotIn-d4 1mg
SDP
Supplier:  Cayman Chemical 356041 MG
Encompass_Preferred
An internal standard for the quantification of N'-nitrosonornicotine by GC-or LC-MS
Catalog No. 50-290-9955