Learn More
Medchemexpress LLC N-nitrososarcosine | 13256-22-9 | MFCD00152671 | 98.0% | 118.09 g/mol | C3H6N2O3 | 25 MG

Supplier: Medchemexpress LLC HYW18817825MG
N-nitrososarcosine is the N-nitroso derivative of sarcosine supplied as a research-grade chemical for analytical, toxicology, and nitrosation studies. It is used as a reference standard and for investigating nitrosamine formation and biological effects.
- Provides a reference for analytical method development and validation.
- Applicable to studies of nitrosation chemistry and nitrosamine toxicology.
- Supplied in milligram quantities suitable for laboratory research.
- Molecular formula C3H6N2O3; molecular weight 118.09 g/mol.
- CAS number 13256-22-9.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.