Learn More
Cayman Chemical N-NornucIferIn 10mg
SDP
Supplier:  Cayman Chemical 3475610 MG
Encompass_Preferred
An alkaloid with cholinesterase inhibitory activity; an inhibitor of butyrylcholinesterase (BChE; IC50 = 5.6 µM)
Catalog No. 50-291-0223