Learn More
Medchemexpress LLC N,O-bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane | 25561-30-2 | MFCD00008269 | 98.0% | 257.40 | C8H18F3NOSi2 | 5G

Supplier: Medchemexpress LLC HY1237985G
N,O-bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent used to derivatize polar functional groups (for example, hydroxyls, amines, and carboxyls) into volatile trimethylsilyl derivatives to improve volatility and thermal stability for gas chromatographic and analytical applications. The formulation includes trimethylchlorosilane to enhance silylation efficiency. For research use only.
- Facilitates formation of trimethylsilyl derivatives for gas chromatography.
- Contains trimethylchlorosilane as an additive to increase silylation efficiency.
- High purity: 98.0%.
- Chemical formula: C8H18F3NOSi2.
- Molecular weight: 257.40.
- Includes supporting documents such as SDS, COA, and data sheet.
- Intended for research and analytical applications only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.