Learn More
Medchemexpress LLC N,O-Dimethylhydroxylamine hydrochloride | 6638-79-5 | >=98.0% | 97.54 | 1 ML
Supplier: Medchemexpress LLC HY4107410MM/1ML
N,O-Dimethylhydroxylamine hydrochloride is a biochemical reagent utilized as an anomeric protecting group in oligosaccharide synthesis, intended exclusively for research applications.
- Used as an anomeric protecting group for oligosaccharide synthesis.