Learn More
Medchemexpress LLC N-octanoyl-L-homoserine lactone | 147852-84-4 | MFCD11113139 | 100.0% | 227.30 g/mol | C12H21NO3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY124237AR50MG
Encompass_Preferred
N-Octanoyl-L-homoserine lactone is a purified analytical standard of the quorum-sensing signaling molecule (CAS 147852-84-4). Provided as a white to off-white solid with high reported purity, it is intended for research use in studies of bacterial communication, analytical method development, and investigations of infection-related pathways.
  • Analytical standard for quorum sensing studies.
  • High purity suitable for analytical and research applications.
  • Available in small pack sizes for laboratory testing.
  • Solid form for straightforward storage and handling.
  • Useful for method development and calibration in analytical workflows.
Catalog No. 50-002-36893