Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 100mg | 29836-26-8 | 292.37 g/mol | C14H28O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116285100MG
Encompass_Preferred
n-Octyl β-D-glucopyranoside is a mild, non-ionic alkyl glucoside detergent used to solubilize, stabilize, and facilitate crystallization of membrane proteins for biochemical and biotechnological research. Supplied as a powder, it dissolves in water and DMSO and is commonly used for protein extraction, reconstitution, and biochemical assays.
  • Non-ionic detergent for solubilization of membrane proteins.
  • High solubility in water and DMSO (100 mg/mL; may require ultrasonic treatment).
  • Supplied as a powder with defined storage stability (powder: -20°C for 3 years; 4°C for 2 years).
  • Low denaturing properties suitable for protein reconstitution and crystallization.
  • Molecular weight 292.37 g/mol; chemical formula C14H28O6.
Catalog No. 50-002-27153