Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 1g | 29836-26-8 | 292.37 g/mol | C14H28O6 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1162851G
Encompass_Preferred
n-Octyl β-D-glucopyranoside is a mild, non-ionic alkyl glucoside detergent used to solubilize and reconstitute membrane-bound proteins for biochemical assays and protein preparation. It is a sugar-based surfactant with molecular formula C14H28O6 and molecular weight 292.37 g/mol.
  • mild, non-denaturing detergent suitable for membrane protein solubilization
  • effective for reconstitution of membrane proteins into liposomes or detergent micelles
  • forms micelles with a critical micelle concentration around 18-20 mM
  • soluble in common organic solvents such as DMSO, DMF, and ethanol
  • supplied as a powdered solid for convenient weighing and storage
Catalog No. 50-002-29469