Learn More
Medchemexpress LLC n-Octyl B-D-glucopyr 500mg | 29836-26-8 | 292.37 g/mol | C14H28O6 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY116285500MG
Encompass_Preferred
n-Octyl β-D-glucopyranoside is a non-ionic alkyl-glycoside detergent used to solubilize and stabilize membrane proteins for biochemical and structural studies. It is supplied as a white to off-white solid and is stored under recommended conditions to preserve purity and performance.
  • Non-ionic detergent for mild, non-denaturing solubilization of membrane proteins.
  • High purity suitable for biochemical and structural applications.
  • Solid, white to off-white appearance for straightforward handling and weighing.
  • Stable under recommended storage conditions to maintain integrity.
  • Soluble in common solvents and DMSO for convenient solution preparation.
Catalog No. 50-002-29873