Learn More
Cayman Chemical n-OctylD-thIoglucopyrnosI 5g
SDP
Supplier:  Cayman Chemical 171845 G
Encompass_Preferred
A low molecular weight nonionic detergent that is commonly used for cell lysis and non-denaturing protein solubilization
Catalog No. 50-296-6454