Learn More
Research Products International Corp n-Octylglucoside, 100 G
SDP
Supplier:  Research Products International Corp N02007100.0
Encompass_Preferred
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Catalog No. 50-197-6840