Learn More
Cayman Chemical n-OctylglucosIde 5g
SDP
Supplier:  Cayman Chemical 143275 G
Encompass_Preferred
Non-denaturing detergent used to solubilize and reconstitute membrane-bound proteins; characterized by a high critical micelle concentration (0.7%) which facilitates ready removal from final protein extracts
Catalog No. 50-295-5444