Learn More
Medchemexpress LLC N-octylnortadalafil | 1173706-35-8 | 98.0% | 487.59 g/mol | C29H33N3O4 | 5 MG

Supplier: Medchemexpress LLC HY1370765MG
N-octylnortadalafil is a synthetic tadalafil derivative supplied as a research reagent for biochemical assays and analytical standards. It is intended for laboratory research use only and is provided in small milligram quantities.
- Intended for research use only.
- Used as a biochemical assay reagent or analytical standard.
- CAS number 1173706-35-8.
- Typical purity reported ~98.0%.
- Molecular formula C29H33N3O4 and molecular weight 487.59 g/mol.
- Supplied in small milligram quantities (example: 5 MG).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.