Learn More
Medchemexpress LLC HY-113204 10mg , N-Oleoyl glycine CAS:2601-90-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11320410MG
Encompass_Preferred
HY-113204 10mg N-Oleoyl glycine CAS: 2601-90-3 N-Oleoyl glycine is a lipoamino acid, which stimulates adipogenesis associated with activation of CB1 receptor and Akt signaling pathway in 3T3-L1 adipocyte. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1670