Learn More
TARGETMOL CHEMICALS INC N-Oleoyl-L-Serine 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T360655MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. N-Oleoyl-L-Serine (N-Oleoylserine) is an endogenous long-chain fatty acid amide that is a lipid modulator of bone remodeling and stimulates osteoclast apoptosis which can be used to study osteoporosis. N-Oleoyl-L-Serine showed high activity in an osteoblast proliferation assay.N-Oleoyl-L-Serine promotes osteoclast apoptosis by inhibiting Erk1/2 phosphorylation and expression of nuclear kB ligand (RANKL) receptor activator in bone marrow stromal cells and osteoblasts, which attenuates osteoclast populations. Purity 97.94%
Catalog No. 50-287-9010