Learn More
TARGETMOL CHEMICALS INC N-Oleoyl Leucine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T357851MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-Oleoyl Leucine (Oleoyl-L-leucine) is an N-fatty acyl amino acid present in plasma. Purity 98.25%
Catalog No. 50-287-9339