Learn More
Medchemexpress LLC N-omega-propyl-L-arginine hydrochloride | 2321366-46-3 | 252.74 g/mol | C9H21ClN4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY102062A100MG
Encompass_Preferred
Nω-Propyl-L-arginine hydrochloride is a research-grade small-molecule inhibitor selective for neuronal nitric oxide synthase (nNOS). Supplied as the hydrochloride salt, it is intended for biochemical and pharmacological research applications to probe nNOS-mediated pathways.
  • Potent, competitive inhibitor of neuronal nitric oxide synthase.
  • Hydrochloride salt form for improved handling.
  • Molecular formula C9H21ClN4O2; molecular weight 252.74 g/mol.
  • Provided as a 100 mg research pack.
  • Intended for in vitro biochemical and pharmacological studies.
Catalog No. 50-002-45499