Learn More
Cayman Chemical N-OxalylglycIn 10mg
SDP
Supplier:  Cayman Chemical 1394410 MG
Encompass_Preferred
A cell permeable inhibitor of α-ketoglutarate-dependent enzymes, including JMJD2A, JMJD2C, and JMJD2E (IC50s = 250, 500, and 24 μM, respectively); inhibits the prolyl hydroxylase domain-containing proteins PHD1 and PHD2 with IC50 values of 2.1 and 5.6 μM, respectively
Catalog No. 50-295-4280