Learn More
TARGETMOL CHEMICALS INC N-OXALYLGLYCINE 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T21634500MG
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. N-Oxalylglycine (Oxalylglycine) is a cell permeable inhibitor of (alpha)-ketoglutarate-dependent enzymes. purity: 98%
Catalog No. 50-258-1016