Learn More
Medchemexpress LLC N-palmitoyl serinol | 126127-31-9 | MFCD05863968 | 99.9% | 329.52 | C19H39NO3 | 10MM 1ML

Supplier: Medchemexpress LLC HY12540710MM 1ML
Palmitoyl serinol (N-palmitoyl serinol) is a bioactive lipid analog used in in vitro research on lipid signaling and epidermal barrier function. It is available as a ready-to-use solution and as solid material for custom preparation.
- CAS number 126127-31-9.
- Molecular formula C19H39NO3.
- Molecular weight 329.52 g/mol.
- Purity 99.89%.
- Offered formulations: 10 mM 1 mL solution in DMSO; solids 5 mg-100 mg.
- Solubility: DMSO 50 mg/mL (151.74 mM); requires ultrasonic or warming to dissolve.
- Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
- Intended for in vitro research applications in lipid biology and skin barrier studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.