Learn More
Medchemexpress LLC N-pentanoyl 2-benzyltryptamine | 220339-00-4 | 99.0% | 334.45 g/mol | C22H26N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10762810MG
Encompass_Preferred
N-Pentanoyl 2-benzyltryptamine is a research compound that acts as a potent and selective antagonist of the MT2 melatonin receptor. It is used in receptor pharmacology to study MT2-mediated signaling and antagonist potency.
  • Potent MT2 antagonist (human MT2 pKi = 8.03).
  • High selectivity versus MT1 and mel1c receptors (~89-fold and ~229-fold).
  • High purity (99.0%).
  • Molecular formula C22H26N2O and molecular weight 334.45 g/mol.
  • Supplied in small-mass quantities suitable for in vitro research (5 mg-100 mg).
  • Useful for antagonist profiling and receptor signaling studies.
Catalog No. 50-002-44523