Learn More
Medchemexpress LLC N-pentanoyl-2-benzyltryptamine | 220339-00-4 | MFCD02262121 | 99.0% | 334.45 g/mol | C22H26N2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1076285MG
Encompass_Preferred
N-Pentanoyl 2-benzyltryptamine is a research-grade small molecule that acts as a selective antagonist of the MT2 melatonin receptor. It is used in receptor pharmacology and melatonin-signaling studies to probe MT2-specific activity. The compound is supplied as a high-purity solid for laboratory use.
  • Selective MT2 melatonin receptor antagonist (human MT2 pKi = 8.03).
  • High purity (99.0%) suitable for research applications.
  • Solid, white to off-white powder for convenient handling.
  • Molecular formula C22H26N2O; molecular weight 334.45 g/mol.
  • Available in small mg quantities for targeted pharmacology studies.
Catalog No. 50-002-42152