Learn More
Cayman Chemical N-PeptIdetrIfluoroACTAte25mg
SDP
Supplier:  Cayman Chemical 3747125 MG
Encompass_Preferred
An E. coli SurA-binding protein; binds to the E. coli periplasmic molecular chaperone SurA (Kds = 2.3 and 1.69 µM for the full-length protein and core domain, respectively)
Catalog No. 50-291-6116