Learn More
Medchemexpress LLC N-Phenethylbenzamide | 3278-14-6 | 99.56% | 225.29 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY32135
Encompass_Preferred
N-Phenethylbenzamide is an active compound extracted from *Liriodendron tulipifera*. It is suitable for the research of inflammatory diseases, demonstrating anti-inflammatory effects by inhibiting LPS-induced NO production in RAW264.7 cells.
  • Active compound
  • Extracted from *Liriodendron tulipifera*
  • Can be used for the research of inflammatory diseases
  • Inhibits LPS-induced NO production with an IC50 value of >50 μM
Catalog No. 50-004-02887