Learn More
Medchemexpress LLC N-phenyl-4-(quinolin-3-ylmethyl)piperidine-1-carboxamide | 959151-50-9 | MFCD10567109 | 99.0% | 345.44 g/mol | C22H23N3O | 10 MG

Supplier: Medchemexpress LLC HY1808110MG
PF 750 is a selective, covalent inhibitor of fatty acid amide hydrolase (FAAH) used in biochemical and cellular research. It shows reported FAAH IC50 values of 16.2-595 nM depending on assay conditions. The compound is supplied as a solid of high purity and is intended for in vitro studies when prepared in appropriate solvents such as DMSO.
- Selective covalent FAAH inhibitor with reported IC50 16.2-595 nM.
- High purity (≈99.0%) suitable for research applications.
- Molecular weight 345.44 g/mol; formula C22H23N3O.
- Soluble in DMSO at 100 mg/mL; ultrasonic treatment recommended.
- Powder storage: -20°C (stable up to 3 years).
- In-solution storage: -80°C for 6 months or -20°C for 1 month.
- Available in small milligram quantities for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.