Learn More
Medchemexpress LLC N-(phenylacetyl-d5)glycine | 1189920-31-7 | MFCD12022247 | ≥99.0% | 198.23 g/mol | C10H6D5NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW015061S1MG
Encompass_Preferred
N-(Phenylacetyl-d5)glycine is the deuterium-labeled isotopologue of phenylacetylglycine supplied as an analytical standard for biochemical and metabolomics research. It is intended for use as an internal standard or tracer in mass spectrometry and quantitative isotope-enrichment studies, offering consistent performance due to high isotopic enrichment and purity.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (≥99.0%) for accurate quantitation.
  • Suitable for metabolic tracing and isotope-enrichment studies.
  • Available in small pack sizes for analytical workflows.
  • Molecular formula C10H6D5NO3; molecular weight 198.23 g/mol.
Catalog No. 50-003-03158