Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Phenylacetyl-L-homoserine lactone | 55154-48-8 | 99.6% | 219.24 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY137407100MG

Encompass_Preferred

N-Phenylacetyl-L-homoserine lactone is presented as a white to off-white solid. This product has been tested and complies with the given specifications.

  • Appearance: White to off-white solid
  • Molecular formula: C₁₂H₁₃NO₃
  • Molecular weight: 219.24
  • Purity (LCMS): 99.60%
  • Storage: -20°C for 3 years (powder); -80°C for 6 months, -20°C for 1 month (in solvent)

Catalog No. 50-003-51328


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.