Learn More
Medchemexpress LLC N-phenylacrylamide | 2210-24-4 | MFCD00080617 | 99.9% | 147.17 g·mol⁻¹ | C9H9NO | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0185825G
Encompass_Preferred
N-Phenylacrylamide (CAS 2210-24-4) is a research-grade monomer used as an ochratoxin A binding material and in mycotoxin extraction and food-safety research. It is provided as a solid powder and as a ready-to-use DMSO solution. Analytical data list a molecular formula of C9H9NO and a molecular weight of 147.17 g·mol⁻¹; high-purity batches are available for analytical and preparative workflows.
  • High affinity for ochratoxin A, enabling selective binding applications.
  • Available as powder and as a 10 mM solution in DMSO for rapid use.
  • High-purity material suitable for analytical assays.
  • Multiple pack sizes accommodate small-scale screening and larger prep work.
  • Useful for mycotoxin extraction and agricultural/food-safety studies.
Catalog No. 50-003-01297