Learn More
Medchemexpress LLC N-phenylpivalamide | 6625-74-7 | MFCD00043691 | 100.0% | 177.24 g/mol | C11H15NO | 10 G

Supplier: Medchemexpress LLC HYW04310510G
N-Phenylpivalamide is an anilide (pivalanilide) used as a chemical building block and drug intermediate for the synthesis of biologically active compounds. It is supplied for research and chemical synthesis applications, typically in small laboratory quantities.
- Synonym: pivalanilide
- Cas number: 6625-74-7
- Molecular formula: C11H15NO
- Molecular weight: 177.24 g/mol
- High purity suitable for synthesis (listed purity 99.98%)
- Available in small lab quantities, including 10 G packaging
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.