Learn More
Medchemexpress LLC N-phenylpivalamide | 6625-74-7 | MFCD00043691 | 100.0% | 177.24 g/mol | C11H15NO | 50 G

Supplier: Medchemexpress LLC HYW04310550G
N-phenylpivalamide (pivalanilide) is a solid organic building block used in organic synthesis and the preparation of drug intermediates. It has a molecular formula of C11H15NO, a molecular weight of 177.24 g/mol, and is supplied in multiple package sizes including 50 g.
- Used as a building block for synthesis of biologically active compounds.
- Solid at room temperature.
- High listed purity (99.98%).
- Available in multiple packaging sizes including 50 g.
- Contains a tert-butyl amide functional group (pivalamide).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.