Learn More
Medchemexpress LLC N-Phthaloyl-β-alanine | 3339-73-9 | 219.20 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0082345G
Encompass_Preferred
N-Phthaloyl-β-alanine is an FX-type hapten without a CH2S moiety. It can be coupled to OVA using the mixed anhydride method and exhibits more heterologies. It is for research use only.
  • FX-type hapten without a CH2S moiety
  • Can be coupled to OVA using the mixed anhydride method
  • Exhibits more heterologies
  • For research use only
Catalog No. 50-004-47996