Learn More
Medchemexpress LLC N-phthaloylglycine | 4702-13-0 | MFCD00005900 | 100.0% | 205.17 | C10H7NO4 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW014665100G
Encompass_Preferred
N-phthaloylglycine is an FX-type hapten that attaches to free amine groups on proteins and is used to derivatize or label proteins for conjugation and immunochemical assays. It is supplied as a high-purity reagent intended for research use only.
  • Attaches covalently to free amine groups on proteins.
  • Suitable for hapten-protein conjugation and immunochemistry applications.
  • Available as solid packs and solution formats.
  • High reported purity suitable for analytical and preparative use.
  • Intended for research use only, not for human use.
Catalog No. 50-003-02751