Learn More
Medchemexpress LLC N-phthaloylglycine | 4702-13-0 | MFCD00005900 | 100.0% | 205.17 g/mol | C10H7NO4 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01466550G
Encompass_Preferred
N-Phthaloylglycine (phthaloyl glycine) is an FX-type hapten used to attach to free amine groups on proteins, enabling haptenization and conjugation workflows for immunochemical and biochemical research. It is supplied as a high-purity solid and as a ready-to-use solution in DMSO to support different experimental needs.
  • High purity (99.95%) solid material.
  • Available as a 10 mM solution in DMSO for direct use.
  • Reacts with free primary amines for protein haptenization.
  • Suitable for immunochemistry and conjugation applications.
  • Offered in multiple solid pack sizes for scale flexibility.
Catalog No. 50-003-03317