Learn More
Medchemexpress LLC N-piperidine Ibrutinib hydrochloride | 2231747-18-3 | 98.1% | 50 MG
Supplier: Medchemexpress LLC HY13098350MG
N-piperidine Ibrutinib hydrochloride is a reversible Ibrutinib derivative and a potent BTK inhibitor. It can be used as a BTK ligand in the synthesis of various PROTACs.
- Potent BTK inhibitor with IC50s of 51.0 nM for WT BTK and 30.7 nM for C481S BTK.
- Can be used as a BTK ligand in the synthesis of PROTACs.
- Enables synthesis of potent PROTAC BTK degraders.